Learn More
Cayman Chemical L-α-Aminoadipic Acid, Purity: ≥95%, CAS: 1118-90-7, 500 mg
SDP
Catalog No. 502965193
Encompass_Preferred
A gliotoxic analog of the excitatory neurotransmitter glutamate that competitively inhibits glutamine synthetase (Ki = 209 µM); actively accumulates in glial cells and inhibits the glutamate transporter with a (Ki = 192 µM)
Catalog No. 50-296-5193 Supplier Cayman Chemical Supplier No. 17579500 MG
A gliotoxic analog of the excitatory neurotransmitter glutamate that competitively inhibits glutamine synthetase (Ki = 209 µM); actively accumulates in glial cells and inhibits the glutamate transporter with a (Ki = 192 µM)