Learn More
Cayman Chemical L-α-Hydroxyglutaric Acid, Purity: ≥95%, CAS: 13095-48-2, 10 mg
SDP
Catalog No. 502982222
Encompass_Preferred
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; inhibits α-ketoglutarate-dependent oxygenases, including several involved in histone lysine and DNA demethylation
Catalog No. 50-298-2222 Supplier Cayman Chemical Supplier No. 2112310 MG
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; inhibits α-ketoglutarate-dependent oxygenases, including several involved in histone lysine and DNA demethylation