Learn More
Cayman Chemical L-a-HydroxyglutarIc AcId 5mg
SDP
Catalog No. 502983333
Encompass_Preferred
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; inhibits α-ketoglutarate-dependent oxygenases, including several involved in histone lysine and DNA demethylation
Catalog No. 50-298-3333 Supplier Cayman Chemical Supplier No. 211235 MG
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; inhibits α-ketoglutarate-dependent oxygenases, including several involved in histone lysine and DNA demethylation