Learn More
Cayman Chemical L-a-HydroxyglutarIc AcIdod 5mg
SDP
Catalog No. 502951985
Encompass_Preferred
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; competitively inhibits α-ketoglutarate-dependent dioxygenases, including several involved in histone lysine and DNA demethylation
Catalog No. 50-295-1985 Supplier Cayman Chemical Supplier No. 118765 MG
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; competitively inhibits α-ketoglutarate-dependent dioxygenases, including several involved in histone lysine and DNA demethylation