Learn More
Selleck Chemical LLC L-a-Phosphatidylcholine S4788-50mg
SDP
Catalog No. 503109199
Encompass_Preferred
L- -phosphatidylcholine a surfactant and a constituent of the liposomal carrier system has shown hepatoprotection in the various hepatotoxicity model
Catalog No. 50-310-9199 Supplier Selleck Chemical LLC Supplier No. S478850MG
L- -phosphatidylcholine a surfactant and a constituent of the liposomal carrier system has shown hepatoprotection in the various hepatotoxicity model