Learn More
Medchemexpress LLC L-a-Phosphatidylserine sodium | 383908-63-2 | 99.0% | 808.06 | 1 MG
SDP
Catalog No. 5000392443
Encompass_Preferred
L-a-Phosphatidylserine sodium is an antigen targeting phosphatidylserine (PS) that induces the production of polyclonal antibodies. It triggers complement-dependent immune damage by binding to PS on the cell membrane surface, activating the complement system. This process causes liposome membrane damage and content release, participating in the regulation of immune response and cell apoptosis signaling. It is used in immunological research.
  • Induces production of polyclonal antibodies
  • Triggers complement-dependent immune damage
  • Activates complement system upon antibody recognition
  • Causes liposome membrane damage and content release
  • Regulates immune response and cell apoptosis signaling
  • Suitable for antibody specificity analysis
  • Suitable for complement activation mechanism research
Catalog No. 50-003-92443 Supplier Medchemexpress LLC Supplier No. HY1650841MG
L-a-Phosphatidylserine sodium is an antigen targeting phosphatidylserine (PS) that induces the production of polyclonal antibodies. It triggers complement-dependent immune damage by binding to PS on the cell membrane surface, activating the complement system. This process causes liposome membrane damage and content release, participating in the regulation of immune response and cell apoptosis signaling. It is used in immunological research.
  • Induces production of polyclonal antibodies
  • Triggers complement-dependent immune damage
  • Activates complement system upon antibody recognition
  • Causes liposome membrane damage and content release
  • Regulates immune response and cell apoptosis signaling
  • Suitable for antibody specificity analysis
  • Suitable for complement activation mechanism research