Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Abrine, TCI America™
SDP

Catalog No. A14891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14891G Supplier TCI America Supplier No. A14891G

missing translation for 'validMainframeMsg'

Specifications

CAS 526-31-8
Color White
Molecular Formula C12H14N2O2
MDL Number MFCD00005645
Quantity 1 g
Synonym Nalpha-Methyl-L-tryptophan
InChI Key CZCIKBSVHDNIDH-NSHDSACASA-N
SMILES CN[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-(methylamino)propanoic acid
Molecular Weight (g/mol) 218.26
PubChem CID 160511
ChEBI CHEBI:15334
Formula Weight 218.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Abrine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.