Learn More
FUJIFILM Biosciences L-Ascorbic Acid Phosphate Magnesium Salt n-Hydrate, 1713265-25-8, MFCD03428049, 10G
Catalog No. NC0276518
Encompass_Preferred
Molecular Formula: (C6H6O9P)・3/2Mg・nH, Molecular Weight: 289.54(anhydride), Grade: Biochemistry, Content: 90.0+% (HPLC), Storage: Room Temperature
Catalog No. NC0276518 Supplier FUJIFILM Biosciences Supplier No. 01312061
Molecular Formula: (C6H6O9P)・3/2Mg・nH, Molecular Weight: 289.54(anhydride), Grade: Biochemistry, Content: 90.0+% (HPLC), Storage: Room Temperature