Learn More
Medchemexpress LLC L-Adenosine | 3080-29-3 | 99.8% | 1 ML
SDP
Catalog No. 5000410684
Encompass_Preferred
L-Adenosine is a metabolically stable enantiomeric analog and a potential probe. It exhibits weakly inhibitory adenosine deaminase (ADA) activity with a Ki value of 385 μM. This compound can be utilized for research into adenosine uptake and accumulation.
  • Metabolically stable enantiomeric analog.
  • Potential probe for research.
  • Weakly inhibits adenosine deaminase activity.
  • Can be used for adenosine uptake research.
  • Can be used for adenosine accumulation research.
Catalog No. 50-004-10684 Supplier Medchemexpress LLC Supplier No. HYD085510MM/1ML
L-Adenosine is a metabolically stable enantiomeric analog and a potential probe. It exhibits weakly inhibitory adenosine deaminase (ADA) activity with a Ki value of 385 μM. This compound can be utilized for research into adenosine uptake and accumulation.
  • Metabolically stable enantiomeric analog.
  • Potential probe for research.
  • Weakly inhibits adenosine deaminase activity.
  • Can be used for adenosine uptake research.
  • Can be used for adenosine accumulation research.