Learn More
Medchemexpress LLC L-adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.24 g/mol | C10H13N5O4 | 50 MG
SDP
Catalog No. 5000247541
Encompass_Preferred
L-adenosine is an enantiomeric analog of adenosine used as a research reagent and a weak adenosine deaminase (ADA) inhibitor (reported Ki = 385 μM). It is supplied as a high-purity solid for studies of adenosine uptake, accumulation, and ADA-related metabolism.
  • High purity - 99.82% (reported).
  • Suitable for adenosine uptake and accumulation studies.
  • Weak ADA inhibitor (Ki ≈ 385 μM).
  • Available as a 50 MG solid; additional pack sizes and DMSO solutions are offered.
  • Molecular weight 267.24 g/mol; formula C10H13N5O4.
Catalog No. 50-002-47541 Supplier Medchemexpress LLC Supplier No. HY10323750MG
L-adenosine is an enantiomeric analog of adenosine used as a research reagent and a weak adenosine deaminase (ADA) inhibitor (reported Ki = 385 μM). It is supplied as a high-purity solid for studies of adenosine uptake, accumulation, and ADA-related metabolism.
  • High purity - 99.82% (reported).
  • Suitable for adenosine uptake and accumulation studies.
  • Weak ADA inhibitor (Ki ≈ 385 μM).
  • Available as a 50 MG solid; additional pack sizes and DMSO solutions are offered.
  • Molecular weight 267.24 g/mol; formula C10H13N5O4.