Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.25 | C10H13N5O4 | 10 MG
SDP

Catalog No. 5000247699
Encompass_Preferred

L-Adenosine is the L-enantiomer of adenosine, a metabolically stable nucleoside analog used as a research probe. It exhibits weak inhibitory activity against adenosine deaminase (ADA) and is used to study adenosine uptake and accumulation.

  • CAS number 3080-29-3.
  • Molecular formula C10H13N5O4; molecular weight 267.25.
  • Purity 99.82% (typical).
  • Physical form solid, white to off-white.
  • Storage: 4°C, protect from light; in solvent -80°C for 6 months, -20°C for 1 month.
  • Reported Ki for ADA: 385 μM.

Catalog No. 50-002-47699 Supplier Medchemexpress LLC Supplier No. HY10323710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Adenosine is the L-enantiomer of adenosine, a metabolically stable nucleoside analog used as a research probe. It exhibits weak inhibitory activity against adenosine deaminase (ADA) and is used to study adenosine uptake and accumulation.

  • CAS number 3080-29-3.
  • Molecular formula C10H13N5O4; molecular weight 267.25.
  • Purity 99.82% (typical).
  • Physical form solid, white to off-white.
  • Storage: 4°C, protect from light; in solvent -80°C for 6 months, -20°C for 1 month.
  • Reported Ki for ADA: 385 μM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.