Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.25 g/mol | C10H13N5O4 | 1 ML
SDP

Catalog No. 5000247822
Encompass_Preferred

L-Adenosine is a metabolically stable enantiomeric analog used as a research probe with weak adenosine deaminase (ADA) inhibitory activity, suitable for studies of adenosine uptake and accumulation.

  • 10 mM solution in DMSO, ready for use
  • Pack size: 1 mL
  • CAS number: 3080-29-3
  • Purity: 99.82%
  • Molecular weight: 267.25 g/mol
  • Store stock solution at -80°C for long-term stability; protect from light

Catalog No. 50-002-47822 Supplier Medchemexpress LLC Supplier No. HY10323710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Adenosine is a metabolically stable enantiomeric analog used as a research probe with weak adenosine deaminase (ADA) inhibitory activity, suitable for studies of adenosine uptake and accumulation.

  • 10 mM solution in DMSO, ready for use
  • Pack size: 1 mL
  • CAS number: 3080-29-3
  • Purity: 99.82%
  • Molecular weight: 267.25 g/mol
  • Store stock solution at -80°C for long-term stability; protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.