Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.25 g/mol | C10H13N5O4 | 100 MG
SDP

Catalog No. 5000247072
Encompass_Preferred

L-Adenosine is an enantiomeric nucleoside used as a biochemical probe for adenosine uptake and metabolism. It displays weak adenosine deaminase (ADA) inhibitory activity and is supplied in high-purity forms for laboratory research.

  • High purity (≈99.8%).
  • Molecular weight 267.25 g/mol.
  • CAS number 3080-29-3.
  • Available as solid and 10 mM DMSO solution formats.
  • Suitable for biochemical, uptake, and metabolism studies.

Catalog No. 50-002-47072 Supplier Medchemexpress LLC Supplier No. HY103237100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Adenosine is an enantiomeric nucleoside used as a biochemical probe for adenosine uptake and metabolism. It displays weak adenosine deaminase (ADA) inhibitory activity and is supplied in high-purity forms for laboratory research.

  • High purity (≈99.8%).
  • Molecular weight 267.25 g/mol.
  • CAS number 3080-29-3.
  • Available as solid and 10 mM DMSO solution formats.
  • Suitable for biochemical, uptake, and metabolism studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.