Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.25 g/mol | C10H13N5O4 | 25 MG
SDP

Catalog No. 5000247173
Encompass_Preferred

L-adenosine is a high-purity adenosine deaminase (ADA) inhibitor provided as a powdered research reagent for in vitro biochemical and cellular studies. The 25 mg vial is suitable for assay development, enzymatic assays, and dose-response experiments, with documented solubility and recommended storage conditions to preserve stability.

  • High purity (99.82%).
  • Suitable for in vitro biochemical and cellular assays.
  • Powdered solid supplied as a 25 mg vial for small-scale studies.
  • Soluble in DMSO (~8.33 mg/mL) with ultrasonic assistance; hygroscopic solvent may affect solubility.
  • Store at 4°C protected from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-47173 Supplier Medchemexpress LLC Supplier No. HY10323725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-adenosine is a high-purity adenosine deaminase (ADA) inhibitor provided as a powdered research reagent for in vitro biochemical and cellular studies. The 25 mg vial is suitable for assay development, enzymatic assays, and dose-response experiments, with documented solubility and recommended storage conditions to preserve stability.

  • High purity (99.82%).
  • Suitable for in vitro biochemical and cellular assays.
  • Powdered solid supplied as a 25 mg vial for small-scale studies.
  • Soluble in DMSO (~8.33 mg/mL) with ultrasonic assistance; hygroscopic solvent may affect solubility.
  • Store at 4°C protected from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.