Learn More
Medchemexpress LLC L-adenosine | 3080-29-3 | MFCD01075784 | 99.8% | 267.25 g/mol | C10H13N5O4 | 5 MG
SDP
Catalog No. 5000247528
Encompass_Preferred
L-Adenosine is the L-enantiomer of adenosine used as a research reagent and probe. It exhibits weak adenosine deaminase (ADA) inhibitory activity and is supplied in high purity for biochemical and pharmacological studies.
  • L-enantiomer of adenosine with defined stereochemistry.
  • Weak adenosine deaminase (ADA) inhibitor suitable for enzyme assays.
  • High reported purity (99.8%).
  • Molecular formula C10H13N5O4 and molecular weight 267.25 g/mol.
  • Available as solid and as pre-made 10 mM DMSO stock for convenient use.
Catalog No. 50-002-47528 Supplier Medchemexpress LLC Supplier No. HY1032375MG
L-Adenosine is the L-enantiomer of adenosine used as a research reagent and probe. It exhibits weak adenosine deaminase (ADA) inhibitory activity and is supplied in high purity for biochemical and pharmacological studies.
  • L-enantiomer of adenosine with defined stereochemistry.
  • Weak adenosine deaminase (ADA) inhibitor suitable for enzyme assays.
  • High reported purity (99.8%).
  • Molecular formula C10H13N5O4 and molecular weight 267.25 g/mol.
  • Available as solid and as pre-made 10 mM DMSO stock for convenient use.