Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alaninamide hydrochloride, 95%

Catalog No. AAH6301914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6301914 25 g
AAH6301906 5 g
AAH6301922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6301914 Supplier Thermo Scientific Chemicals Supplier No. H6301914
  • $168.00
    $141.65 / Each of 1

    Save $26.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33208-99-0 | C3H9ClN2O | 124.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33208-99-0
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.57
MDL Number MFCD00066145
InChI Key FIAINKIUSZGVGX-DKWTVANSSA-N
Synonym l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride
PubChem CID 2775816
IUPAC Name (2S)-2-aminopropanamide;hydrochloride
SMILES Cl.C[C@H](N)C(N)=O

Specifications

Melting Point 212°C to 217°C (decomposition)
Quantity 25 g
Formula Weight 124.57
Percent Purity 95%
Physical Form Solid
Chemical Name or Material L-Alaninamide hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.