Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Alaninamide, L-phenylalanyl-L-prolyl-N-(4-nitrophenyl)- | 201738-99-0 | 99.19% | 453.49 | 10 MG
SDP

Catalog No. 5000396674
Encompass_Preferred

Phe-Pro-Ala-pNA is a chromogenic substrate of tripeptidyl peptidase. It can be used to test tripeptidyl peptidase activity.

  • Chromogenic substrate of tripeptidyl peptidase
  • Used to test tripeptidyl peptidase activity
  • Purity of 99.19%
  • Molecular formula: C23H27N5O5
  • Molecular weight: 453.49
  • Appearance: White to off-white solid

Catalog No. 50-003-96674 Supplier Medchemexpress LLC Supplier No. HYP447010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phe-Pro-Ala-pNA is a chromogenic substrate of tripeptidyl peptidase. It can be used to test tripeptidyl peptidase activity.

  • Chromogenic substrate of tripeptidyl peptidase
  • Used to test tripeptidyl peptidase activity
  • Purity of 99.19%
  • Molecular formula: C23H27N5O5
  • Molecular weight: 453.49
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.