Learn More
Bioworld L-Alanine, 250 g
Catalog No. 50255279
Encompass_Preferred
Utilized in clinical nutritionUsed as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy.Used for cell culture.Utilized to enrich nutrition in health foods and drinks in the food industry.Synonym: (S)-2-Aminopropionic acid, L-α-Aminopropionic acid Technical Information:High Purity GradeSpecific rotation: +13.7º to +15.1ºAppearance: White crystalline powderBoiling Point : 212.9 °CDensity : 1.432 g/cm3HS Code : 29224995IUPAC_Name: AlanineMelting Point: 314.5 °CRTECS Number: AY2990000
Catalog No. 50-255-279 Supplier Bioworld Supplier No. 401002642
Utilized in clinical nutritionUsed as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy.Used for cell culture.Utilized to enrich nutrition in health foods and drinks in the food industry.Synonym: (S)-2-Aminopropionic acid, L-α-Aminopropionic acid Technical Information:High Purity GradeSpecific rotation: +13.7º to +15.1ºAppearance: White crystalline powderBoiling Point : 212.9 °CDensity : 1.432 g/cm3HS Code : 29224995IUPAC_Name: AlanineMelting Point: 314.5 °CRTECS Number: AY2990000