Learn More
Medchemexpress LLC L-alanine-13C2,15N | 312623-85-1 | MFCD34676307 | 99.0% | 92.07 g/mol | C[13C]2H7[15N]O2 | 10 MG
SDP
Catalog No. 5000296328
Encompass_Preferred
L-Alanine-13C2,15N is L-alanine labeled with two 13C atoms and one 15N atom, supplied as a high-purity isotopically labeled standard for use in analytical and metabolic studies.
  • Isotopically labeled with two 13C and one 15N
  • High purity: 99.0%
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS
  • Molecular weight: 92.07 g/mol
  • Available as a 10 MG vial for analytical use
Catalog No. 50-002-96328 Supplier Medchemexpress LLC Supplier No. HYN0229S1510MG
L-Alanine-13C2,15N is L-alanine labeled with two 13C atoms and one 15N atom, supplied as a high-purity isotopically labeled standard for use in analytical and metabolic studies.
  • Isotopically labeled with two 13C and one 15N
  • High purity: 99.0%
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS
  • Molecular weight: 92.07 g/mol
  • Available as a 10 MG vial for analytical use