Learn More
Medchemexpress LLC L-alanine-2-d1 | 21386-65-2 | MFCD00084105 | ≥98.0% | 90.10 g·mol⁻1 | C3H6DNO2 | 250 MG
SDP
Catalog No. 5000282360
Encompass_Preferred
L-Alanine-2-d1 is the deuterium-labeled form of the amino acid L-alanine, labeled at the 2-position. Supplied as a small-quantity solid (commonly 250 mg), it is used as an isotopic tracer, internal standard, or reagent in NMR, mass spectrometry, and metabolic studies.
  • Deuterium label at the 2-position for unambiguous isotopic tracing.
  • High isotopic enrichment (≥98 atom % D) for sensitive analytical work.
  • High chemical purity (≥98%) suitable for standards and synthesis.
  • Suitable as an internal standard for MS and NMR quantification.
  • Stable, easy-to-handle solid for routine laboratory use.
Catalog No. 50-002-82360 Supplier Medchemexpress LLC Supplier No. HYW141857250MG
L-Alanine-2-d1 is the deuterium-labeled form of the amino acid L-alanine, labeled at the 2-position. Supplied as a small-quantity solid (commonly 250 mg), it is used as an isotopic tracer, internal standard, or reagent in NMR, mass spectrometry, and metabolic studies.
  • Deuterium label at the 2-position for unambiguous isotopic tracing.
  • High isotopic enrichment (≥98 atom % D) for sensitive analytical work.
  • High chemical purity (≥98%) suitable for standards and synthesis.
  • Suitable as an internal standard for MS and NMR quantification.
  • Stable, easy-to-handle solid for routine laboratory use.