Learn More
Medchemexpress LLC L-Alanine-3,3,3-d3, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 225101-67-7 | 99.4% | 314.35 | 10 MG
SDP
Catalog No. 5000366938
Encompass_Preferred
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3 and is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66938 Supplier Medchemexpress LLC Supplier No. HYW009204S110MG
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3 and is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.