Learn More
Medchemexpress LLC L-Alanine-3,3,3-d3, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 225101-67-7 | 99.4% | 314.35 | 25 MG
SDP
Catalog No. 5000366735
Encompass_Preferred
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3. It is for research use only and not sold to patients. This product can be used as a tracer and an internal standard for quantitative analysis by various methods. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-66735 Supplier Medchemexpress LLC Supplier No. 25MGHYW009204S1
Fmoc-Ala-OH-3,3,3-d3 is the deuterium labeled Fmoc-Ala-OH-3,3,3. It is for research use only and not sold to patients. This product can be used as a tracer and an internal standard for quantitative analysis by various methods. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs