Learn More
Medchemexpress LLC L-alanine, 3-sulfino-, hydrate (1:1) | 207121-48-0 | MFCD00150732 | 99.8% | 171.17 g/mol | C3H9NO5S | 1 ML
SDP
Catalog No. 5000299771
Encompass_Preferred
L-Cysteinesulfinic acid monohydrate (CAS 207121-48-0) is a research-grade amino acid derivative and metabotropic glutamate receptor (mGluR) agonist supplied for biochemical and pharmacological studies. It is offered as a solid or a ready-to-use aqueous solution to support receptor pharmacology, assay development, and screening.
  • Available as solid or 10 mM solution (1 mL) in water.
  • High purity: 99.8%.
  • Molecular weight 171.17 g/mol; formula C3H9NO5S.
  • Suitable for receptor pharmacology and biochemical assays.
  • Packaged in small sizes for immediate use and screening.
Catalog No. 50-002-99771 Supplier Medchemexpress LLC Supplier No. HYW01723010MM/1ML
L-Cysteinesulfinic acid monohydrate (CAS 207121-48-0) is a research-grade amino acid derivative and metabotropic glutamate receptor (mGluR) agonist supplied for biochemical and pharmacological studies. It is offered as a solid or a ready-to-use aqueous solution to support receptor pharmacology, assay development, and screening.
  • Available as solid or 10 mM solution (1 mL) in water.
  • High purity: 99.8%.
  • Molecular weight 171.17 g/mol; formula C3H9NO5S.
  • Suitable for receptor pharmacology and biochemical assays.
  • Packaged in small sizes for immediate use and screening.