Learn More
Medchemexpress LLC L-alanine | 56-41-7 | MFCD00064410 | 99.9% | 89.09 | C3H7NO2 | 10 MG
SDP
Catalog No. 5000296285
Encompass_Preferred
L-Alanine analytical standard intended for research and analytical applications. Provided as a white solid, this reference material is used for calibration, method validation, and identification in chromatographic and mass spectrometric workflows.
  • Analytical standard for L-alanine
  • High purity (99.9% by assay/HPLC)
  • Chemical formula C3H7NO2; molecular weight 89.09
  • White solid suitable for analytical workflows
  • Provided in a brown glass bottle for stability
  • Useful for chromatography, mass spectrometry, and reference material applications
Catalog No. 50-002-96285 Supplier Medchemexpress LLC Supplier No. HYN0229R10MG
L-Alanine analytical standard intended for research and analytical applications. Provided as a white solid, this reference material is used for calibration, method validation, and identification in chromatographic and mass spectrometric workflows.
  • Analytical standard for L-alanine
  • High purity (99.9% by assay/HPLC)
  • Chemical formula C3H7NO2; molecular weight 89.09
  • White solid suitable for analytical workflows
  • Provided in a brown glass bottle for stability
  • Useful for chromatography, mass spectrometry, and reference material applications