Learn More
Medchemexpress LLC L-Alanine-d7 | 74280-71-0 | 99.8% | 96.14 | 5 MG
SDP
Catalog No. 5000431048
Encompass_Preferred
L-Alanine-d7 is the deuterium-labeled version of L-Alanine. L-Alanine is a non-essential amino acid that plays a role in sugar and acid metabolism, boosts immunity, and provides energy for muscle tissue, the brain, and the central nervous system. This compound is used for research purposes only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-31048 Supplier Medchemexpress LLC Supplier No. HYN0229S135MG
L-Alanine-d7 is the deuterium-labeled version of L-Alanine. L-Alanine is a non-essential amino acid that plays a role in sugar and acid metabolism, boosts immunity, and provides energy for muscle tissue, the brain, and the central nervous system. This compound is used for research purposes only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.