Learn More
Medchemexpress LLC L-Alanine, ethyl ester, hydrochloride (1:1) | 1115-59-9 | 98.0% | 153.61 | 500 G
SDP
Catalog No. 5000429231
Encompass_Preferred
H-Ala-OEt.HCl is an alanine derivative. Amino acids and amino acid derivatives are widely utilized as ergogenic supplements. They play a crucial role in influencing the secretion of anabolic hormones, optimizing fuel supply during exercise, enhancing mental performance in stress-related tasks, and preventing exercise-induced muscle damage. These compounds are recognized as beneficial dietary substances for their ergogenic properties.
  • Alanine derivative
  • Influences anabolic hormone secretion
  • Optimizes fuel supply during exercise
  • Enhances mental performance
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-29231 Supplier Medchemexpress LLC Supplier No. HYI0517500G
H-Ala-OEt.HCl is an alanine derivative. Amino acids and amino acid derivatives are widely utilized as ergogenic supplements. They play a crucial role in influencing the secretion of anabolic hormones, optimizing fuel supply during exercise, enhancing mental performance in stress-related tasks, and preventing exercise-induced muscle damage. These compounds are recognized as beneficial dietary substances for their ergogenic properties.
  • Alanine derivative
  • Influences anabolic hormone secretion
  • Optimizes fuel supply during exercise
  • Enhances mental performance
  • Prevents exercise-induced muscle damage