Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine, MP Biomedicals™

Catalog No. MP21002871 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21002871 100 g
MP21002875 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21002871 Supplier MP Biomedicals, Inc Supplier No. 02100287.1
Add to Cart
Add to Cart

Plays a key role in the glucose-alanine cycle between muscle tissue and the liver. L-Alanine, MP Biomedicals is a nonessential amino acid. In amino acid-degrading tissues, such as muscle, amino groups are pooled as glutamate by transamination reactions.

The amino group of glutamate is transferred to pyruvate via alanine aminotransferase, forming alanine and α-ketoglutarate. The alanine is passed into the blood and transported to the liver. This reaction is reversed in the liver, where pyruvate can be used in gluconeogenesis to form glucose, which may return to other tissues through the circulatory system. Increased alanine levels correlate with higher blood pressure, energy intake, cholesterol levels, and body mass index.

Alanine has been used as an amino donor in a study of the conversion of ketomethiobutyrate to methionine in several species of Bacillus. L-Alanine has been shown to enhance the rates of germination of Bacillus subtilis spores.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Solubility Information Freely soluble in water; sparingly soluble in alcohol; insoluble in ether.
Quantity 100 g
Formula Weight 89.09
Percent Purity ≥99%
Physical Form Crystalline Powder
Recommended Storage Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.