Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-alanine, n-[(1,1-dimethylethoxy)carbonyl]-3-[(1,1-dimethylethoxy)carbonyl]amino | 88971-40-8 | >95.0% | 304.34 | C13H24N2O6 | 5 G
SDP

Catalog No. 5000210353
Encompass_Preferred

Boc-Dap(Boc)-OH is a Boc-protected derivative of 2,3-diaminopropionic acid supplied for research use as an amino-acid building block for peptide and small-molecule synthesis.

  • Boc-protected amino acid building block
  • Suitable for peptide synthesis and related organic synthesis
  • Available in multiple sizes, including 5 G
  • Purity ≥95.0%
  • Molecular weight 304.34 g/mol
  • CAS number 88971-40-8

Catalog No. 50-002-10353 Supplier Medchemexpress LLC Supplier No. HYW0477995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Dap(Boc)-OH is a Boc-protected derivative of 2,3-diaminopropionic acid supplied for research use as an amino-acid building block for peptide and small-molecule synthesis.

  • Boc-protected amino acid building block
  • Suitable for peptide synthesis and related organic synthesis
  • Available in multiple sizes, including 5 G
  • Purity ≥95.0%
  • Molecular weight 304.34 g/mol
  • CAS number 88971-40-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.