Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Alanine, N-[(4-methylphenyl)sulfonyl]-, 5-phenyl-1H-pyrrol-3-yl ester | 99740-00-8 | 99.7% | 100 MG
SDP

Catalog No. 5000410410
Encompass_Preferred

5-Phenyl-1H-pyrrol-3-yl tosyl-L-alaninate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular formula: C20H20N2O4S
  • Molecular weight: 384.45
  • Purity: 99.7%
  • Target: Biochemical assay reagents
  • Storage conditions: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month), protect from light

Catalog No. 50-004-10410 Supplier Medchemexpress LLC Supplier No. HYD0864100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Phenyl-1H-pyrrol-3-yl tosyl-L-alaninate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular formula: C20H20N2O4S
  • Molecular weight: 384.45
  • Purity: 99.7%
  • Target: Biochemical assay reagents
  • Storage conditions: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month), protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.