Learn More
Medchemexpress LLC L-Alanine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-N-(phenylmethyl)- | 672917-68-9 | 99.6% | 401.45 | C25H23NO4 | 25 MG
SDP
Catalog No. 5000208765
Encompass_Preferred
N-Fmoc-N-benzyl-L-alanine is an Fmoc-protected N-benzyl derivative of L-alanine used as a protected amino acid building block in peptide synthesis and organic chemistry research. It is supplied as an off-white to light yellow solid with high reported purity and stability when stored cold.
  • Fmoc-protected amino acid building block.
  • High purity suitable for synthetic applications.
  • Available as a powder; store refrigerated or frozen for long-term stability.
  • Off-white to light yellow solid appearance.
  • Commonly used in solid-phase peptide synthesis.
Catalog No. 50-002-08765 Supplier Medchemexpress LLC Supplier No. 25MGHYW072732
N-Fmoc-N-benzyl-L-alanine is an Fmoc-protected N-benzyl derivative of L-alanine used as a protected amino acid building block in peptide synthesis and organic chemistry research. It is supplied as an off-white to light yellow solid with high reported purity and stability when stored cold.
  • Fmoc-protected amino acid building block.
  • High purity suitable for synthetic applications.
  • Available as a powder; store refrigerated or frozen for long-term stability.
  • Off-white to light yellow solid appearance.
  • Commonly used in solid-phase peptide synthesis.