Learn More
Medchemexpress LLC L-alanine N,N,O-d3 | 19470-97-4 | MFCD06658795 | 99.0% | 92.11 | C3H4D3NO2 | 10 MG
SDP
Catalog No. 5000296355
Encompass_Preferred
L-Alanine-d3-1 is the deuterium-labeled analogue of L-alanine used as a stable isotope tracer and internal standard for quantitative analytical methods. It is supplied as a high-purity isotopically labeled amino acid for use in NMR, GC-MS, and LC-MS assays, method development, and metabolic studies.
  • Deuterium-labeled L-alanine (N,N,O-d3) for isotope tracing and standards.
  • High purity suitable for quantitative analysis (99.0%).
  • Compatible with NMR, GC-MS, and LC-MS applications.
  • Useful as an internal standard for accurate quantitation.
  • Provided in small research-scale quantities for method development.
Catalog No. 50-002-96355 Supplier Medchemexpress LLC Supplier No. HYN0229S1610MG
L-Alanine-d3-1 is the deuterium-labeled analogue of L-alanine used as a stable isotope tracer and internal standard for quantitative analytical methods. It is supplied as a high-purity isotopically labeled amino acid for use in NMR, GC-MS, and LC-MS assays, method development, and metabolic studies.
  • Deuterium-labeled L-alanine (N,N,O-d3) for isotope tracing and standards.
  • High purity suitable for quantitative analysis (99.0%).
  • Compatible with NMR, GC-MS, and LC-MS applications.
  • Useful as an internal standard for accurate quantitation.
  • Provided in small research-scale quantities for method development.