Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Alanine, N2-acetyl-L-arginyl-L-phenylalanyl-L-alanyl-L-alanyl-L-lysyl-L-alanyl- (TFA) | 99.6% | 775.90 | 10 MG
SDP

Catalog No. 5000422777
Encompass_Preferred

Ac-RFAAKAA-COOH (TFA) is a heptapeptide identified as L-Alanine, N2-acetyl-L-arginyl-L-phenylalanyl-L-alanyl-L-alanyl-L-lysyl-L-alanyl- (TFA). This white solid has an LCMS purity of 99.62% and a molecular weight of 775.90 (free base).

  • Appears as a white solid
  • Purity of 99.62% (LCMS)
  • Molecular weight of 775.90 (free base)
  • Nitrogen content: 16.35%
  • Peptide content: 82.41%
  • Tested and complies with given specifications
  • Sealed storage required, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • For research use only, not fully validated for medical applications

Catalog No. 50-004-22777 Supplier Medchemexpress LLC Supplier No. HYP10940A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-RFAAKAA-COOH (TFA) is a heptapeptide identified as L-Alanine, N2-acetyl-L-arginyl-L-phenylalanyl-L-alanyl-L-alanyl-L-lysyl-L-alanyl- (TFA). This white solid has an LCMS purity of 99.62% and a molecular weight of 775.90 (free base).

  • Appears as a white solid
  • Purity of 99.62% (LCMS)
  • Molecular weight of 775.90 (free base)
  • Nitrogen content: 16.35%
  • Peptide content: 82.41%
  • Tested and complies with given specifications
  • Sealed storage required, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • For research use only, not fully validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.