Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine, Spectrum™ Chemical
SDP

Catalog No. 18620173 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-173 25 kg
18-620-169 25 g
18-620-170 100 g
18-620-171 1 kg
18-620-172 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-173 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL11525KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56-41-7 | 89.09 g/mol

Chemical Identifiers

CAS 56-41-7

Specifications

Color White
Packaging Poly Bottle
Molecular Weight (g/mol) 89.09
Quantity 25 kg
Percent Purity 98.5 to 102%
Grade Reagent
Chemical Name or Material L-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.