Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine USP/EP/JP/FCC, Ajinomoto

Catalog No. 502946020
Encompass
Change view
Click to view available options
Quantity:
1 kg
10 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-294-6020 50 kg
50-294-6061 1 kg
50-294-6009 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6020 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A20111
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-41-7
Molecular Weight (g/mol) 89.09

Specifications

Packaging Fiber Drum
Quantity 50 kg
Percent Purity 98.5 to 101.0%
Chemical Name or Material L-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.