Learn More
Research Products International Corp L-Alanine, USP Grade, 100 Grams
SDP
Supplier:  Research Products International Corp A20060100.0
Encompass_Preferred
L-Alanine is a crucial building block for protein synthesis. It is incorporated into polypeptide chains during translation, contributing to the structure and function of proteins in various biological processes. L-Alanine serves as a precursor for the biosynthesis of other important molecules, including nucleotides and certain coenzymes. 
L-Alanine is used in cell culture media in laboratories for the cultivation of cells. Its presence contributes to the overall metabolic pathways within cells and is essential for supporting the growth and viability of cells in artificial environments.
Catalog No. 50-213-119