Learn More
Medchemexpress LLC HY-16933 1mg Medchemexpress, L-Alanosine CAS:5854-93-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY169331MG
Encompass_Preferred
Medchemexpress, HY-16933 1mg L-Alanosine CAS:5854-93-3 L-Alanosine (NSC-153353), an antibiotic from Streptomyces alanosinicus, has antineoplastic activity. L-Alanosine (NSC-153353) inhibits adenylosuccinate synthetase , which converts inosine monophospate (IMP) into adenylosuccinate. L-Alanosine (NSC-153353) blocks the common de novo purine biosynthesis pathway and, thereby, inhibits tumor cells with MTAP deficiency [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2786