Learn More
Medchemexpress LLC L-Alanyl-(4R)-4-hydroxy-L-proline | 76400-25-4 | 99.4% | 202.21 | 10 MG
SDP
Catalog No. 5000398318
Encompass_Preferred
H-Ala-Hyp-OH, also known as L-Alanyl-(4R)-4-hydroxy-L-proline, is a biologically active peptide supplied as a white to off-white solid. It is intended for research use only.
  • Biologically active peptide
  • White to off-white solid appearance
  • Soluble in DMSO at 100 mg/mL
  • Store powder at -80°C for up to 2 years or -20°C for up to 1 year
Catalog No. 50-003-98318 Supplier Medchemexpress LLC Supplier No. HYP533810MG
H-Ala-Hyp-OH, also known as L-Alanyl-(4R)-4-hydroxy-L-proline, is a biologically active peptide supplied as a white to off-white solid. It is intended for research use only.
  • Biologically active peptide
  • White to off-white solid appearance
  • Soluble in DMSO at 100 mg/mL
  • Store powder at -80°C for up to 2 years or -20°C for up to 1 year