Learn More
Medchemexpress LLC HY-W014102 100mg Medchemexpress, L-Alanyl-L-glutamine CAS:39537-23-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW014102100MG
Encompass_Preferred
Medchemexpress, HY-W014102 100mg L-Alanyl-L-glutamine CAS:39537-23-0 L-Alanyl-L-glutamine, a glutamine dipeptide, is benefit for the antioxidant system, attenuating inflammation, and may modulate the heat shock protein (HSP) response in catabolic situations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3356