Learn More
AdipoGen L-Alanyl-L-glutamine
SDP
Catalog No. 502561787
Encompass_Preferred
Chemical. CAS 39537-23-0. Formula C8H15N3O4. MW 217.22. Synthetic. L-Glutamine is an essential supplement for cell culture. The free amino acid L-glutamine is unstable and degrades to pyroglutamate. Alanyl-L-glutamine Ala-Gln and Gylcyl-L-glutamine Gly-Gln are used in cell culture media as alternative delivery forms of stabilized L-glutamine. Alanyl-glutamine is a widely used alternative supplement to L-glutamine in the production of biopharmaceuticals, cosmetics and food products. Alanyl-glutamine also acts as an antioxidant peroxide and anti-apoptosis LPS-induced factor. Ala-Gln has been used in studies on injury and sepsis, and on the effects of irradiation on leucine and protein metabolism in vivo. Ala-Gln has been utilized to investigate polymorphonuclear leucocyte and myeloperoxidase activity in vitro. Shown to improve intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy.
Catalog No. 50-256-1787 Supplier AdipoGen Supplier No. CDXA0222G010
Chemical. CAS 39537-23-0. Formula C8H15N3O4. MW 217.22. Synthetic. L-Glutamine is an essential supplement for cell culture. The free amino acid L-glutamine is unstable and degrades to pyroglutamate. Alanyl-L-glutamine Ala-Gln and Gylcyl-L-glutamine Gly-Gln are used in cell culture media as alternative delivery forms of stabilized L-glutamine. Alanyl-glutamine is a widely used alternative supplement to L-glutamine in the production of biopharmaceuticals, cosmetics and food products. Alanyl-glutamine also acts as an antioxidant peroxide and anti-apoptosis LPS-induced factor. Ala-Gln has been used in studies on injury and sepsis, and on the effects of irradiation on leucine and protein metabolism in vivo. Ala-Gln has been utilized to investigate polymorphonuclear leucocyte and myeloperoxidase activity in vitro. Shown to improve intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy.