Learn More
Apexbio Technology LLC L-Alanyl-L-Glutamine 39537-23-0 25g
SDP
Catalog No. 5000087624
Encompass_Preferred
L-Alanyl-L-glutamine (CAS 39537-23-0) is a stable water-soluble dipeptide consisting of L-glutamine and L-alanine Following oral or enteral administration it is hydrolyzed in the gastrointestinal tract locally supporting maintenance of the intestinal mucosal barrier and modulating barrier function This activity results in reduced bacterial translocation and a lowered risk of infection and associated complications such as diarrhea and malabsorption The dipeptide enhances intestinal uptake of L-glutamine compared to free L-glutamine and has been shown to influence antioxidant capacity attenuate inflammatory responses and regulate heat shock protein expression under catabolic conditions L-Alanyl-L-glutamine serves as a versatile tool for studies of gut physiology nutrient absorption and cellular stress responses in biomedical research
Catalog No. 50-000-87624 Supplier Apexbio Technology LLC Supplier No. B822825000
L-Alanyl-L-glutamine (CAS 39537-23-0) is a stable water-soluble dipeptide consisting of L-glutamine and L-alanine Following oral or enteral administration it is hydrolyzed in the gastrointestinal tract locally supporting maintenance of the intestinal mucosal barrier and modulating barrier function This activity results in reduced bacterial translocation and a lowered risk of infection and associated complications such as diarrhea and malabsorption The dipeptide enhances intestinal uptake of L-glutamine compared to free L-glutamine and has been shown to influence antioxidant capacity attenuate inflammatory responses and regulate heat shock protein expression under catabolic conditions L-Alanyl-L-glutamine serves as a versatile tool for studies of gut physiology nutrient absorption and cellular stress responses in biomedical research