Learn More
Medchemexpress LLC L-Alanyl-L-glutamine | 39537-23-0 | MFCD00133046 | 95.0% | 217.22 g/mol | C8H15N3O4 | 1 ML
SDP
Catalog No. 5000304030
Encompass_Preferred
L-Alanyl-L-glutamine is a stable dipeptide of alanine and glutamine supplied as a 10 mM aqueous solution for research use. It provides a soluble, longer-lasting source of glutamine for cell culture and biochemical assays, and is reported to support antioxidant defenses, attenuate inflammation, and modulate heat shock protein responses in catabolic conditions.
  • Stable glutamine source for cell culture applications.
  • Ready-to-use 10 mM aqueous solution for convenience and reproducibility.
  • Supports antioxidant defenses and attenuates inflammatory responses.
  • May modulate heat shock protein (HSP) pathways in stressed cells.
  • Helps maintain mitochondrial metabolism and secretion coupling in β-cells.
  • High purity suitable for research and analytical use.
Catalog No. 50-003-04030 Supplier Medchemexpress LLC Supplier No. HYW01410210MM/1ML
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
L-Alanyl-L-glutamine is a stable dipeptide of alanine and glutamine supplied as a 10 mM aqueous solution for research use. It provides a soluble, longer-lasting source of glutamine for cell culture and biochemical assays, and is reported to support antioxidant defenses, attenuate inflammation, and modulate heat shock protein responses in catabolic conditions.
  • Stable glutamine source for cell culture applications.
  • Ready-to-use 10 mM aqueous solution for convenience and reproducibility.
  • Supports antioxidant defenses and attenuates inflammatory responses.
  • May modulate heat shock protein (HSP) pathways in stressed cells.
  • Helps maintain mitochondrial metabolism and secretion coupling in β-cells.
  • High purity suitable for research and analytical use.