Learn More
ATHENA ES L-Alanyl-L-Glutamine, dehydrated
Supplier:  ATHENA ES 045325
Encompass_Preferred
L-Alanyl-L-glutamine is a direct replacement for L-glutamine in cell culture media. The alanyl derivative of glutamine is thermally stable, which extends the shelf-life of cell culture medium by limiting deamination of the terminal amino group of the side-chain.
Catalog No. 50-001-58145