Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanyl-L-tryptophan 98.0+%, TCI America™
SDP

Catalog No. A12621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12621G Supplier TCI America Supplier No. A12621G

Chemical Identifiers

CAS 16305-75-2
Molecular Formula C14H17N3O3
Molecular Weight (g/mol) 275.308
MDL Number MFCD00037948
InChI Key WUGMRIBZSVSJNP-UFBFGSQYSA-N
Synonym H-Ala-Trp-OH
PubChem CID 85362
ChEBI CHEBI:73808
IUPAC Name (2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-(1H-indol-3-yl)propanoic acid
SMILES CC(C(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O)N

Specifications

Color White
Quantity 1 g
Formula Weight 275.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanyl-L-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.