Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanyl-L-tyrosine 99.0+%, TCI America™
SDP

Catalog No. A12631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12631G Supplier TCI America Supplier No. A12631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3061-88-9
Molecular Formula C12H16N2O4
Molecular Weight (g/mol) 252.27
MDL Number MFCD00038164
InChI Key ALZVPLKYDKJKQU-XVKPBYJWSA-N
Synonym H-Ala-Tyr-OH
PubChem CID 92946
ChEBI CHEBI:73395
IUPAC Name (2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N

Specifications

Color White
Quantity 1 g
Formula Weight 252.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanyl-L-tyrosine
EINECSNumber (3)-4664
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.