Learn More
Medchemexpress LLC L-albizziin | 1483-07-4 | 98.0% | 5 MG
SDP
Catalog No. 5000230223
Encompass_Preferred
L-albizziin is a non-proteinogenic amino acid that functions as a sulfhydryl group reagent and a glutamase inhibitor. It is used primarily in vitro for studies of glutamase activity and for cancer research.
  • Sulfhydryl group reagent used in biochemical assays.
  • Glutamase inhibitor for in vitro research applications.
  • High purity: 98.0%.
  • White to off-white solid, stable under recommended storage conditions.
  • Available in small milligram packages suitable for laboratory studies.
Catalog No. 50-002-30223 Supplier Medchemexpress LLC Supplier No. HY1211675MG
L-albizziin is a non-proteinogenic amino acid that functions as a sulfhydryl group reagent and a glutamase inhibitor. It is used primarily in vitro for studies of glutamase activity and for cancer research.
  • Sulfhydryl group reagent used in biochemical assays.
  • Glutamase inhibitor for in vitro research applications.
  • High purity: 98.0%.
  • White to off-white solid, stable under recommended storage conditions.
  • Available in small milligram packages suitable for laboratory studies.