Learn More
Medchemexpress LLC L-alloisoleucine-d10 | 202529-06-4 | 141.23 | C6H3D10NO2 | 10 MG
SDP
Catalog No. 5000297612
Encompass_Preferred
L-alloisoleucine-d10 is the deuterium-labeled (d10) isotopologue of L-alloisoleucine supplied as an analytical reference standard for isotope-dilution and mass spectrometry applications. It retains the stereochemistry of L-alloisoleucine while providing a mass-shifted internal standard for accurate quantitation in biological and analytical workflows.
  • Deuterium labeled isotopologue for use as an internal standard.
  • Suitable for mass spectrometry and isotope-dilution assays.
  • Supplied in small, accurately weighed packs for analytical use.
  • Stable under recommended storage; consult certificate of analysis for purity and handling.
Catalog No. 50-002-97612 Supplier Medchemexpress LLC Supplier No. HYI1060S10MG
L-alloisoleucine-d10 is the deuterium-labeled (d10) isotopologue of L-alloisoleucine supplied as an analytical reference standard for isotope-dilution and mass spectrometry applications. It retains the stereochemistry of L-alloisoleucine while providing a mass-shifted internal standard for accurate quantitation in biological and analytical workflows.
  • Deuterium labeled isotopologue for use as an internal standard.
  • Suitable for mass spectrometry and isotope-dilution assays.
  • Supplied in small, accurately weighed packs for analytical use.
  • Stable under recommended storage; consult certificate of analysis for purity and handling.