Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Allothreonine | 28954-12-3 | 119.12 | 500 MG
SDP

Catalog No. 5000446149
Encompass_Preferred

L-Allothreonine (H-allo-Thr-OH) is an endogenous metabolite primarily used for research purposes. This compound is not intended for patient use.

  • Solid, white to off-white appearance
  • Molecular formula: C4H9NO3
  • Melting point: 272°C
  • Boiling point: 345.8°C at 760 mmHg
  • Relative density: 1.3126 g/cm³
  • Soluble in water (12.5 mg/mL)
  • Tested for stereochemistry effects on L1210 mouse leukemia cell line in vitro

Catalog No. 50-004-46149 Supplier Medchemexpress LLC Supplier No. HYW008315500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Allothreonine (H-allo-Thr-OH) is an endogenous metabolite primarily used for research purposes. This compound is not intended for patient use.

  • Solid, white to off-white appearance
  • Molecular formula: C4H9NO3
  • Melting point: 272°C
  • Boiling point: 345.8°C at 760 mmHg
  • Relative density: 1.3126 g/cm³
  • Soluble in water (12.5 mg/mL)
  • Tested for stereochemistry effects on L1210 mouse leukemia cell line in vitro

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.