Learn More
Apexbio Technology LLC L-Allothreonine 28954-12-3 200mg
SDP
Catalog No. 5000088414
Encompass_Preferred
L-Allothreonine (CAS 28954-12-3) is a stereoisomer of threonine and is observed as an endogenous metabolite in mammalian and microbial metabolic pathways It participates in amino acid metabolism influencing both protein synthesis and degradation processes L-Allothreonine is frequently employed in metabolomic studies to assess alterations in amino acid metabolic pathways under disease conditions and to elucidate regulatory mechanisms Additionally it serves as a substrate in in vitro metabolic models to characterize amino acid interactions contributing to research on cellular and systemic metabolism
Catalog No. 50-000-88414 Supplier Apexbio Technology LLC Supplier No. C6711200
L-Allothreonine (CAS 28954-12-3) is a stereoisomer of threonine and is observed as an endogenous metabolite in mammalian and microbial metabolic pathways It participates in amino acid metabolism influencing both protein synthesis and degradation processes L-Allothreonine is frequently employed in metabolomic studies to assess alterations in amino acid metabolic pathways under disease conditions and to elucidate regulatory mechanisms Additionally it serves as a substrate in in vitro metabolic models to characterize amino acid interactions contributing to research on cellular and systemic metabolism