Learn More
Medchemexpress LLC L-Allylglycine | 16338-48-0 | >=98.0% | 115.13 | 50 G
SDP
Catalog No. 5000454130
Encompass_Preferred
L-Allylglycine is an amino acid derivative and an inhibitor for glutamate decarboxylase (GAD). It reduces GABA biosynthesis in the brain and exhibits convulsant activity. This product is intended for research use only.
  • Inhibits glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity
  • Soluble in H2O (50 mg/mL, requires ultrasonic)
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Reduced GABA content in hippocampus, cortex, and cerebellum in vivo
  • Induced running fits, partial rigidity, and convulsions in rats
Catalog No. 50-004-54130 Supplier Medchemexpress LLC Supplier No. HYW01290650G
L-Allylglycine is an amino acid derivative and an inhibitor for glutamate decarboxylase (GAD). It reduces GABA biosynthesis in the brain and exhibits convulsant activity. This product is intended for research use only.
  • Inhibits glutamate decarboxylase (GAD)
  • Reduces GABA biosynthesis in the brain
  • Exhibits convulsant activity
  • Soluble in H2O (50 mg/mL, requires ultrasonic)
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Reduced GABA content in hippocampus, cortex, and cerebellum in vivo
  • Induced running fits, partial rigidity, and convulsions in rats