Learn More
Medchemexpress LLC L-allylglycine hydrochloride | 195316-72-4 | MFCD06797042 | 98.0% | 151.59 g/mol | C5H10ClNO2 | 1 G
SDP
Catalog No. 5000300729
Encompass_Preferred
L-allylglycine hydrochloride is the hydrochloride salt of L-allylglycine, an amino acid derivative that functions as a glutamate decarboxylase (GAD) inhibitor. It reduces GABA biosynthesis and is used in neuroscience research to model GABAergic disruption and convulsant activity.
  • Inhibits glutamate decarboxylase (GAD), decreasing GABA production.
  • Useful as a research tool to study GABAergic pathways and seizure mechanisms.
  • Provided as a hydrochloride salt for improved solubility in aqueous media.
  • Available in small research-scale quantities suitable for experimental use.
  • Characterized by formula C5H10ClNO2 and molecular weight 151.59 g/mol.
Catalog No. 50-003-00729 Supplier Medchemexpress LLC Supplier No. HYW0155951G
L-allylglycine hydrochloride is the hydrochloride salt of L-allylglycine, an amino acid derivative that functions as a glutamate decarboxylase (GAD) inhibitor. It reduces GABA biosynthesis and is used in neuroscience research to model GABAergic disruption and convulsant activity.
  • Inhibits glutamate decarboxylase (GAD), decreasing GABA production.
  • Useful as a research tool to study GABAergic pathways and seizure mechanisms.
  • Provided as a hydrochloride salt for improved solubility in aqueous media.
  • Available in small research-scale quantities suitable for experimental use.
  • Characterized by formula C5H10ClNO2 and molecular weight 151.59 g/mol.